Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021472.4 | NP_067447.2 | 2 | 410 | ||
NM_201239.3 | NP_957691.1 | 2 | 707 | |||
GenBank mRNA | AB041044.1 | - | 2 | 316 | ||
AK050451.1 | - | 2 | 307 | |||
AK076504.1 | - | 2 | 380 | |||
AY762362.1 | - | 2 | 309 | |||
BC005569.1 | - | 2 | 284 | 3967265 | ||
BC061105.1 | - | 2 | 291 | 30296907 | ||
BC066843.1 | - | 2 | 371 | 30059308 |