Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 5, 6 | 657 | ||
GenBank mRNA | AF263247.1 | - | 5, 6 | 619 | ||
AK007307.1 | - | 5, 6 | 628 | |||
AK034859.1 | - | 5, 6 | 656 | |||
AK049424.1 | - | 5, 6 | 677 | |||
AK075637.1 | - | 4, 5 | 497 | |||
AK088854.1 | - | 5, 6 | 665 | |||
AK145800.1 | - | 5, 6 | 720 | |||
AK152523.1 | - | 5, 6 | 665 | |||
AK168788.1 | - | 5, 6 | 683 | |||
AK169149.1 | - | 5, 6 | 665 | |||
BC039054.1 | - | 5, 6 | 643 | 2648667 | ||
BC058687.1 | - | 5, 6 | 655 | 6411707 |