Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021895.2 | NP_068695.1 | 2 | 283 | ||
GenBank mRNA | AJ289242.2 | - | 2 | 283 | ||
AK145322.1 | - | 2 | 336 | |||
AK150044.1 | - | 2 | 285 | |||
BC013616.1 | - | 2 | 292 | 3157024 | ||
BC027032.1 | - | 2 | 234 | |||
BC087554.1 | - | 2 | 236 | 5351470 | ||
DQ303411.1 | - | 2 | 186 |