Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022022.3 | NP_071305.2 | 20 | 3077 | ||
GenBank mRNA | AF260926.1 | - | 20 | 3192 | ||
AK029914.1 | - | 20 | 2682 | |||
AK083409.1 | - | 20 | 2680 | |||
AK122345.1 | - | 20 | 2588 | |||
AK163319.1 | - | 8 | 874 | |||
BC067402.1 | - | 20 | 2824 | 30099547 | ||
BC075620.1 | - | 20 | 3038 | 6405514 |