Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025793.3 | NP_080069.2 | 9, 10 | 1076 | ||
GenBank mRNA | AK002352.1 | - | 9, 10 | 1037 | ||
AK013170.1 | - | 9, 10 | 1026 | |||
AK146101.1 | - | 9, 10 | 1077 | |||
AK151975.1 | - | 9, 10 | 1038 | |||
AK168603.1 | - | 9, 10 | 1029 | |||
AK170602.1 | - | 9, 10 | 1023 | |||
BC004595.1 | - | 9, 10 | 1020 | 3584839 |