Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164290.1 | NP_001157762.1 | 10 | 933 | ||
NM_001164291.1 | NP_001157763.1 | 9 | 825 | |||
NM_001164292.1 | NP_001157764.1 | 9 | 822 | |||
NM_001164293.1 | NP_001157765.1 | 9 | 792 | |||
NM_001164294.1 | NP_001157766.1 | 9 | 771 | |||
NM_001164295.1 | NP_001157767.1 | 9 | 768 | |||
NM_024192.2 | NP_077154.2 | 10 | 924 | |||
GenBank mRNA | AB232359.1 | - | 8 | 723 | ||
AK003457.1 | - | 9 | 727 | |||
AK013884.1 | - | 9 | 783 | |||
BC003442.1 | - | 10 | 888 | 2650719 |