Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290416.1 | NP_001277345.1 | 17, 18 | 1908 | ||
NM_026146.4 | NP_080422.1 | 17, 18 | 1814 | |||
GenBank mRNA | AK019490.2 | - | 17, 18 | 1808 | ||
AY074931.1 | - | 16, 17 | 1751 | |||
BC027043.1 | - | 17, 18 | 2094 |