Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 8 | 1148 | ||
GenBank mRNA | AK006731.1 | - | 1343 | |||
AK007202.1 | - | 8 | 1130 | |||
AK015594.1 | - | 9 | 1359 | |||
AK016626.1 | - | 1153 | ||||
AK137605.1 | - | 3 | 454 | |||
BC030908.1 | - | 6 | 768 | |||
BC051060.1 | - | 4 | 543 | |||
BC051150.1 | - | 8 | 1148 | 6772814 |