Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026340.3 | NP_080616.1 | 4 | 451 | ||
NM_152894.2 | NP_690854.1 | 4 | 451 | |||
GenBank mRNA | AK016547.1 | - | 4 | 438 | ||
AK018297.1 | - | 4 | 420 | |||
BC034742.1 | - | 4 | 430 | 4935194 |