Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam96b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026753.2 | NP_081029.1 | 1, 2 | 173 | ||
GenBank mRNA | AK003830.1 | - | 1, 2 | 176 | ||
BC055880.1 | - | 1, 2 | 147 | 4190754 |