Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271388.1 | NP_001258317.1 | 7 | 1280 | ||
NM_001271389.1 | NP_001258318.1 | 7 | 1280 | |||
NM_001271390.1 | NP_001258319.1 | 6 | 898 | |||
NM_001271391.1 | NP_001258320.1 | 6 | 898 | |||
NM_001271392.1 | NP_001258321.1 | 6 | 898 | |||
NM_023472.2 | NP_075961.1 | 7 | 1280 | |||
GenBank mRNA | AB221648.1 | - | 6 | 783 | ||
AF314031.1 | - | 7 | 1114 | |||
AK003438.2 | - | 6 | 817 | |||
AK054037.1 | - | 7 | 1086 | |||
AK081695.1 | - | 6 | 783 | |||
AK139343.1 | - | 7 | 1162 | |||
AK161446.1 | - | 7 | 1286 | |||
BC037437.1 | - | 6 | 826 | |||
BC066113.1 | - | 6 | 817 | 30361246 | ||
BC079615.1 | - | 7 | 1097 | 30361702 | ||
BF730624.1 | - | 3 | 634 |