Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024205.2 | NP_001019376.1 | 2 | 869 | ||
GenBank mRNA | AK122494.1 | - | 2 | 853 | ||
AK132603.1 | - | 837 | ||||
AK142954.1 | - | 2 | 839 | |||
AK152336.1 | - | 2 | 840 | |||
AK160601.1 | - | 2 | 870 | |||
AK170764.1 | - | 2 | 867 |