Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024205.2 | NP_001019376.1 | 2 | 744 | ||
GenBank mRNA | AK122494.1 | - | 2 | 728 | ||
AK132603.1 | - | 712 | ||||
AK142954.1 | - | 2 | 714 | |||
AK152336.1 | - | 2 | 715 | |||
AK160601.1 | - | 2 | 745 | |||
AK170764.1 | - | 2 | 742 |