Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 2 | 124 | ||
NM_001287211.1 | NP_001274140.1 | 2 | 124 | |||
NM_026972.5 | NP_081248.4 | 2 | 124 | |||
GenBank mRNA | AF373409.1 | - | 2 | 102 | ||
AF374471.1 | - | 2 | 81 | |||
AF422108.1 | - | 2 | 97 | |||
AF424790.1 | - | 2 | 96 | |||
AF424791.1 | - | 2 | 96 | |||
AF424792.1 | - | 2 | 96 | |||
AF424793.1 | - | 2 | 96 | |||
AF424794.1 | - | 2 | 116 | |||
AF424795.1 | - | 2 | 113 | |||
AF424796.1 | - | 2 | 107 | |||
AF424797.1 | - | 2 | 96 | |||
AF424798.1 | - | 2 | 116 | |||
AF424799.1 | - | 2 | 96 | |||
AF424803.1 | - | 2 | 132 | |||
AK007656.1 | - | 1 | 40 | |||
AK138999.1 | - | 2 | 98 |