Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 18, 19 | 2092 | ||
GenBank mRNA | AK004875.2 | - | 18, 19 | 2048 | ||
AK016520.1 | - | 18, 19 | 2062 | |||
AK028589.1 | - | 18, 19 | 2061 | |||
AK144693.1 | - | 18, 19 | 2066 | |||
AK160597.1 | - | 18, 19 | 2077 | |||
AK220287.1 | - | 18, 19 | 2067 | |||
BC025902.1 | - | 10, 11 | 1158 | |||
BC026779.1 | - | 18, 19 | 1993 | 4018298 | ||
BC027428.1 | - | 18, 19 | 2055 | 3983461 |