Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 4 | 546 | ||
GenBank mRNA | AK008493.1 | - | 4 | 580 | ||
AK048210.1 | - | 4 | 546 | |||
AK084881.1 | - | 4 | 580 | |||
AK088600.1 | - | 4 | 589 | |||
AK147765.1 | - | 4 | 579 | |||
AK162563.1 | - | 4 | 579 | |||
AK165916.1 | - | 4 | 608 | |||
AK166237.1 | - | 4 | 632 | |||
BC016240.1 | - | 4 | 564 | 4506688 |