Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143679.1 | - | 8 | 1675 | ||
AK143680.1 | - | 1370 | ||||
AK148140.1 | - | 8 | 1244 | |||
AK151565.1 | - | 8 | 1257 | |||
AK167896.1 | - | 8 | 1235 | |||
AK168069.1 | - | 8 | 1271 | |||
AK186237.1 | - | 1 | 24 | |||
AK201112.1 | - | 1 | 24 | |||
AK204573.1 | - | 1 | 24 | |||
AY424290.1 | - | 8 | 1246 | |||
BC014875.1 | - | 8 | 1246 | 2811173 |