Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028472.2 | NP_082748.1 | 11 | 1629 | ||
GenBank mRNA | AF454954.1 | - | 11 | 1629 | ||
AK014221.1 | - | 9 | 834 | |||
AY263358.1 | - | 11 | 1150 | |||
BC066153.1 | - | 11 | 1134 | 30098453 |