Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028500.3 | NP_082776.2 | 7 | 926 | ||
NM_029782.3 | NP_084058.2 | 5 | 627 | |||
GenBank mRNA | AB206786.1 | - | 7 | 831 | ||
AK006582.1 | - | 7 | 927 | |||
AK046081.1 | - | 5 | 628 |