Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103156.2 | NP_001096626.1 | 6 | 1586 | ||
NM_001103157.2 | NP_001096627.1 | 6 | 1941 | |||
NM_001285469.1 | NP_001272398.1 | 6 | 1495 | |||
NM_001285470.1 | NP_001272399.1 | 5 | 1460 | |||
NM_028734.5 | NP_083010.2 | 6 | 1643 | |||
GenBank mRNA | AK015015.1 | - | 3 | 613 | ||
AK052981.1 | - | 6 | 1645 | |||
AK086787.1 | - | 6 | 1614 | |||
AK145468.1 | - | 6 | 1533 | |||
AK162343.1 | - | 5 | 1422 | |||
AK165644.1 | - | 6 | 1906 | |||
AK166607.1 | - | 6 | 1532 | |||
BC043459.1 | - | 6 | 1496 | |||
BC150881.1 | - | 6 | 1502 | 9087792 | ||
BC150889.1 | - | 6 | 1502 | 9087800 | ||
BC150997.1 | - | 1553 | 9087910 | |||
BC151008.1 | - | 1553 | 9087921 |