Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103156.2 | NP_001096626.1 | 4 | 1210 | ||
NM_001103157.2 | NP_001096627.1 | 4 | 1565 | |||
NM_001285469.1 | NP_001272398.1 | 4 | 1119 | |||
NM_001285470.1 | NP_001272399.1 | 3 | 1084 | |||
NM_001285471.1 | NP_001272400.1 | 3 | 1084 | |||
NM_028734.5 | NP_083010.2 | 4 | 1267 | |||
GenBank mRNA | AK052981.1 | - | 4 | 1269 | ||
AK086787.1 | - | 4 | 1238 | |||
AK089927.1 | - | 3 | 1058 | |||
AK145468.1 | - | 4 | 1156 | |||
AK162343.1 | - | 3 | 1046 | |||
AK165644.1 | - | 4 | 1530 | |||
AK166162.1 | - | 4 | 1234 | |||
AK166607.1 | - | 4 | 1156 | |||
BC043459.1 | - | 4 | 1120 | |||
BC150881.1 | - | 4 | 1126 | 9087792 | ||
BC150889.1 | - | 4 | 1126 | 9087800 | ||
BC150997.1 | - | 1177 | 9087910 | |||
BC151008.1 | - | 1177 | 9087921 |