Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029578.3 | NP_083854.3 | 9 | 723 | ||
GenBank mRNA | AK011439.1 | - | 8 | 611 | ||
AK011555.1 | - | 9 | 714 | |||
AK154341.1 | - | 9 | 700 | |||
AK165685.1 | - | 8 | 606 | |||
AK165955.1 | - | 9 | 707 | |||
BC021419.1 | - | 9 | 699 | 5012877 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK133340.1 | - | 1983 | Chr. - - [-] on Build |