Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001980.2 | NP_001001980.2 | 20 | 2586 | ||
NM_001256122.1 | NP_001243051.1 | 15 | 2348 | |||
GenBank mRNA | AK019457.1 | - | 14 | 2380 | ||
AK122443.1 | - | 13 | 1701 | |||
AK134095.1 | - | 15 | 2149 | |||
AK147547.1 | - | 15 | 2349 | |||
BC042189.1 | - | 12 | 1665 |