Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030081.2 | NP_084357.2 | 12 | 2234 | ||
GenBank mRNA | BC031135.1 | - | 11 | 2065 | 4971294 | |
BC049106.1 | - | 12 | 2234 | 6412839 |