Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033587.3 | NP_291065.3 | 1 | 1091 | ||
GenBank mRNA | AY013798.1 | - | 1 | 1091 | ||
BC049814.1 | - | 1 | 1217 | 6508140 |