Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033588.4 | NP_291066.1 | 1 | 2377 | ||
GenBank mRNA | AY013799.1 | - | 1 | 2377 | ||
BC057404.1 | - | 1 | 2477 | 6405582 |