Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175681.3 | NP_783612.2 | 6 | 907 | ||
GenBank mRNA | AY605231.1 | - | 6 | 626 | ||
BC044746.2 | - | 6 | 907 | 5363416 |
Set Membership: |
G-protein coupled receptor |