Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037754.3 | NP_001032843.1 | 8 | 842 | ||
NM_001037755.3 | NP_001032844.2 | 8 | 842 | |||
NM_130862.4 | NP_570932.2 | 8 | 842 | |||
GenBank mRNA | AF390178.1 | - | 8 | 800 | ||
AF390179.1 | - | 8 | 800 | |||
AK049469.1 | - | 8 | 843 | |||
AK145924.1 | - | 8 | 843 | |||
AK158579.1 | - | 8 | 867 | |||
AK160401.1 | - | 8 | 843 | |||
AK205197.1 | - | 2 | 206 | |||
BC006620.1 | - | 8 | 833 | 3158004 | ||
BC016411.1 | - | 8 | 833 | 3158004 |