Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam212b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163356.1 | NP_001156828.1 | 2 | 903 | ||
NM_175398.4 | NP_780607.2 | 2 | 1050 | |||
GenBank mRNA | AK033558.1 | - | 2 | 983 | ||
AK078368.1 | - | 2 | 903 | |||
BC052371.1 | - | 2 | 983 | 6491043 |