Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003685.3 | NP_001003685.2 | 6 | 582 | ||
GenBank mRNA | AK030504.1 | - | 6 | 536 | ||
AK133683.1 | - | 5 | 1438 | |||
BC120748.1 | - | 6 | 512 | 40129671 | ||
BC120774.1 | - | 6 | 512 | 40129697 | ||
BC144892.1 | - | 6 | 512 | |||
L07379.1 | - | 6 | 515 |
Set Membership: |
G-protein coupled receptor |