Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bspry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138653.1 | NP_619594.1 | 6 | 946 | ||
GenBank mRNA | AJ276690.1 | - | 6 | 946 | ||
AK035588.1 | - | 6 | 952 | |||
AK160930.1 | - | 6 | 952 | |||
BC003992.1 | - | 4 | 489 | |||
BC139079.1 | - | 6 | 962 | 8862101 | ||
BC139080.1 | - | 6 | 962 | 8862102 |