Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011610.3 | NP_035740.2 | 6 | 782 | ||
GenBank mRNA | AK004752.1 | - | 6 | 783 | ||
AK004844.2 | - | 6 | 782 | |||
AK140332.1 | - | 6 | 783 | |||
AK153301.1 | - | 6 | 783 | |||
AK155385.1 | - | 6 | 783 | |||
BC141405.1 | - | 6 | 769 | 9056018 | ||
M59378.1 | - | 6 | 755 | |||
M60469.1 | - | 6 | 753 | |||
X76401.1 | - | 6 | 668 |