Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206924.2 | NP_996807.1 | 1 | 423 | ||
GenBank mRNA | AB016490.1 | - | 1 | 368 | ||
AK004469.1 | - | 1 | 335 | |||
AK089048.1 | - | 1 | 335 | |||
AK146337.1 | - | 1 | 424 | |||
BC008139.1 | - | 1 | 387 | 3582930 |