Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025291.3 | NP_079567.2 | 4 | 459 | ||
GenBank mRNA | AK007302.1 | - | 4 | 445 | ||
AK010375.1 | - | 4 | 445 | |||
AK154232.1 | - | 4 | 429 | |||
BC026480.1 | - | 2 | 238 | |||
BC048362.1 | - | 4 | 431 | 2646489 | ||
BQ748094.1 | - | 2 | 127 |