Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290528.1 | NP_001277457.1 | 4 | 1706 | ||
GenBank mRNA | BC046514.1 | - | 1 | 102 | ||
BC057197.1 | - | 3 | 415 | |||
BC078445.1 | - | 4 | 745 |