Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013797.5 | NP_038825.1 | 2 | 428 | ||
GenBank mRNA | AB031813.1 | - | 2 | 269 | ||
AF148218.1 | - | 2 | 252 | |||
AF223067.1 | - | 2 | 262 | |||
AK041125.1 | - | 2 | 276 | |||
AK165360.1 | - | 2 | 283 | |||
AY195868.1 | - | 2 | 261 | |||
AY195869.1 | - | 2 | 261 | |||
AY243579.1 | - | 2 | 261 | |||
BC041147.1 | - | 2 | 211 | 4975986 |