Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177242.4 | NP_796216.2 | 4 | 903 | ||
GenBank mRNA | AK078914.1 | - | 4 | 911 | ||
AK083481.1 | - | 4 | 920 | |||
AK135118.1 | - | 4 | 937 | |||
AK162261.1 | - | 4 | 467 | |||
AK194078.1 | - | 2 | 101 | |||
BC038622.1 | - | 4 | 543 | |||
BC068149.1 | - | 4 | 903 | 30537082 |