Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026123.3 | NP_080399.1 | 5 | 917 | ||
GenBank mRNA | AK003273.1 | - | 5 | 692 | ||
AK004433.1 | - | 5 | 681 | |||
AK132757.1 | - | 5 | 918 | |||
AK135885.1 | - | 5 | 909 | |||
AK210936.1 | - | 2 | 95 | |||
AK215289.1 | - | 1 | 20 | |||
AY017214.1 | - | 5 | 902 | |||
BC019484.1 | - | 5 | 884 | 4039903 |