Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159635.1 | NP_001153107.1 | 5 | 500 | ||
NM_026434.4 | NP_080710.1 | 5 | 590 | |||
GenBank mRNA | AK008110.1 | - | 5 | 446 | ||
AK021243.1 | - | 5 | 471 | |||
AK035348.1 | - | 5 | 498 | |||
AK036252.1 | - | 5 | 471 | |||
AK135109.1 | - | 4 | 371 | |||
BC005540.1 | - | 5 | 484 | 3590164 | ||
CB183820.1 | - | 5 | 661 |