Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168244.1 | NP_001161716.1 | 3 | 764 | ||
NM_001168246.1 | NP_001161718.1 | 4 | 463 | |||
NM_027954.3 | NP_082230.2 | 4 | 445 | |||
GenBank mRNA | AK005954.1 | - | 4 | 446 | ||
AK167332.1 | - | 3 | 763 | |||
BC094628.1 | - | 4 | 340 | 3374002 | ||
BC132534.1 | - | 4 | 395 | 40130811 | ||
BY083329.1 | - | 3 | 177 | |||
BY246600.1 | - | 4 | 378 |