Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028707.4 | NP_082983.3 | 10 | 1996 | ||
GenBank mRNA | AK018116.1 | - | 5 | 452 | ||
AK034575.1 | - | 10 | 1765 | |||
AK045529.1 | - | 10 | 1727 | |||
AK053835.1 | - | 10 | 1724 | |||
AK149027.1 | - | 10 | 1726 | |||
BC056437.1 | - | 6 | 495 | |||
BC062930.1 | - | 9 | 1577 | 6855293 | ||
BC065053.1 | - | 10 | 1764 | 5709231 | ||
BC065055.1 | - | 10 | 1747 | 5720949 | ||
BC066026.1 | - | 10 | 1996 | 5721455 |