Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175369.3 | NP_780578.1 | 4 | 441 | ||
GenBank mRNA | AK077161.1 | - | 4 | 442 | ||
BC138881.1 | - | 4 | 349 | 8861903 | ||
BC138883.1 | - | 4 | 349 | 8861905 | ||
BC145455.1 | - | 5 | 391 |