Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301649.1 | NP_001288578.1 | 10 | 1285 | ||
NM_007517.4 | NP_031543.2 | 11 | 1436 | |||
GenBank mRNA | AK154707.1 | - | 11 | 1305 | ||
AK158035.1 | - | 11 | 1227 | |||
AK167335.1 | - | 11 | 1288 | |||
AK171011.1 | - | 11 | 1239 | |||
AK171184.1 | - | 11 | 1232 | |||
AK171202.1 | - | 11 | 1246 | |||
AK171249.1 | - | 11 | 1226 | |||
AK171541.1 | - | 11 | 1226 | |||
AK171621.1 | - | 11 | 1219 | |||
AK178251.1 | - | 1 | 54 | |||
BC012699.1 | - | 10 | 1069 | 3986517 | ||
BC016485.1 | - | 11 | 1229 | 3962982 | ||
BM900175.1 | - | 2 | 220 | |||
U41736.1 | - | 11 | 1234 |