Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010829.2 | NP_034959.2 | 4 | 625 | ||
GenBank mRNA | AK011426.1 | - | 4 | 609 | ||
AK040616.1 | - | 3 | 410 | |||
AK078467.1 | - | 4 | 576 | |||
AK083388.1 | - | 4 | 530 | |||
AK133907.1 | - | 4 | 618 | |||
AK161089.1 | - | 4 | 620 | |||
BC150759.1 | - | 4 | 544 | 9087670 | ||
M24919.1 | - | 4 | 908 | |||
M80360.1 | - | 4 | 622 |