Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024622.2 | NP_001019793.1 | 2 | 283 | ||
GenBank mRNA | AK145195.1 | - | 2 | 242 | ||
AK150595.1 | - | 2 | 244 | |||
AK152279.1 | - | 2 | 246 | |||
AK159201.1 | - | 2 | 254 | |||
AK166071.1 | - | 2 | 267 | |||
AK168723.1 | - | 2 | 211 | |||
AK172325.1 | - | 1 | 288 | |||
AK189299.1 | - | 1 | 372 | |||
BC023320.1 | - | 2 | 254 | |||
BC061235.1 | - | 2 | 247 | 6593866 | ||
BC106192.1 | - | 2 | 192 | |||
BC132588.1 | - | 2 | 305 | 40130865 | ||
BC132590.1 | - | 2 | 305 | 40130867 | ||
BC145194.1 | - | 2 | 253 | |||
BY287179.1 | - | 2 | 283 |