Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015789.3 | NP_056604.2 | 5 | 791 | ||
GenBank mRNA | AF177399.1 | - | 5 | 696 | ||
AF274312.1 | - | 5 | 750 | |||
AK131694.1 | - | 5 | 736 | |||
BC002215.1 | - | 5 | 769 | 3490482 | ||
BE134426.1 | - | 1 | 113 |