Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021028.3 | NP_066356.3 | 10 | 805 | ||
GenBank mRNA | AF105217.1 | - | 10 | 780 | ||
AJ249341.2 | - | 10 | 783 | |||
AK041651.1 | - | 10 | 796 | |||
AK051286.1 | - | 9 | 725 | |||
AK087557.1 | - | 10 | 806 | |||
AK154160.1 | - | 10 | 799 | |||
BC019982.1 | - | 10 | 797 | 4159742 |