Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jagn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026365.3 | NP_080641.1 | 1, 2 | 218 | ||
GenBank mRNA | AK003673.1 | - | 1, 2 | 144 | ||
AK017953.1 | - | 1, 2 | 153 | |||
AK141417.1 | - | 1, 2 | 198 | |||
AK151252.1 | - | 1, 2 | 180 | |||
BC011296.1 | - | 1, 2 | 185 | 3962988 | ||
BC025910.1 | - | 1, 2 | 172 | 5252891 | ||
BC037019.1 | - | 1, 2 | 172 | 5364053 | ||
BU609459.1 | - | 1, 2 | 219 |