Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285931.1 | NP_001272860.1 | 1, 2 | 310 | ||
GenBank mRNA | BC058370.1 | - | 1, 2 | 138 | ||
CB525602.1 | - | 1, 2 | 277 |