Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173047.3 | NP_766635.1 | 3 | 911 | ||
GenBank mRNA | AK003232.1 | - | 3 | 917 | ||
AK028150.1 | - | 3 | 887 | |||
BC028763.1 | - | 3 | 921 | 1313642 | ||
BC087735.1 | - | 3 | 872 | 5369996 | ||
BC096658.1 | - | 3 | 881 | 5145435 |